Skip to Content
Merck
All Photos(1)

Key Documents

589128

Sigma-Aldrich

2-Ethylacrylic acid

contains 150 ppm BHT as inhibitor, 98%

Synonym(s):

2-Ethylpropenoic acid, 2-Methylenebutanoic acid, 2-Methylenebutyric acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€332.00

€332.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€332.00

About This Item

Linear Formula:
H2C=C(C2H5)CO2H
CAS Number:
Molecular Weight:
100.12
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€332.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

contains

150 ppm BHT as inhibitor

refractive index

n20/D 1.437 (lit.)

bp

176 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

[H]\C([H])=C(\CC)C(O)=O

InChI

1S/C5H8O2/c1-3-4(2)5(6)7/h2-3H2,1H3,(H,6,7)

InChI key

WROUWQQRXUBECT-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

181.9 °F - closed cup

Flash Point(C)

83.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O A Mamer et al.
The Biochemical journal, 160(3), 417-426 (1976-12-15)
1. Normal human urine contains small amounts (less than 4 mg/g of creatinine) of 2-ethylhydracrylic acid, formed, we believe, by a previously undisclosed endogenous catabolic pathway for the oxidation of a newly described series of R metabolites of isoleucine. 2.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service