Skip to Content
Merck
All Photos(1)

Key Documents

576271

Sigma-Aldrich

7-Aminoflavone

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€29.10

€29.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€29.10

About This Item

Empirical Formula (Hill Notation):
C15H11NO2
CAS Number:
Molecular Weight:
237.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€29.10


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

184-188 °C (lit.)

functional group

ketone
phenyl

SMILES string

Nc1ccc2C(=O)C=C(Oc2c1)c3ccccc3

InChI

1S/C15H11NO2/c16-11-6-7-12-13(17)9-14(18-15(12)8-11)10-4-2-1-3-5-10/h1-9H,16H2

InChI key

JGSKYGCPGCBEET-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Stompor et al.
Journal of agricultural and food chemistry, 63(30), 6749-6756 (2015-07-16)
Xanthohumol (14) and isoxanthohumol (6) derived from hop (Humulus lupulus L., Cannabaceae) and selected chalcone and chromene derivatives, obtained by chemical synthesis, were studied for antifeedant activity against the peach-potato aphid (Myzus persicae [Sulz.]). The study used also commercially available

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service