Skip to Content
Merck
All Photos(1)

Key Documents

576263

Sigma-Aldrich

6-Aminoflavone

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€32.40

€32.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€32.40

About This Item

Empirical Formula (Hill Notation):
C15H11NO2
CAS Number:
Molecular Weight:
237.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€32.40


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

196-200 °C (lit.)

functional group

ketone
phenyl

SMILES string

Nc1ccc2OC(=CC(=O)c2c1)c3ccccc3

InChI

1S/C15H11NO2/c16-11-6-7-14-12(8-11)13(17)9-15(18-14)10-4-2-1-3-5-10/h1-9H,16H2

InChI key

YWHKBWPNCFDUPQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Almeqdad Y Habashneh et al.
Archiv der Pharmazie, 347(6), 415-422 (2014-03-13)
A new series of N1-(flavon-6-yl)amidrazones were synthesized by reacting the hydrazonoyl chloride derived from 6-aminoflavone with the appropriate sec-cyclic amines. The antitumor activities of these compounds were evaluated on breast cancer (MCF-7) and leukemic (K562) cell lines. Among the compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service