Skip to Content
Merck
All Photos(2)

Key Documents

574090

Sigma-Aldrich

Gallium(III) perchlorate hydrate

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€282.00

€282.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€282.00

About This Item

Linear Formula:
Ga(CIO4)3 · xH2o
CAS Number:
Molecular Weight:
368.07 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
chunks

€282.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.999% trace metals basis

form

chunks

impurities

≤20.0 ppm Trace Metal Analysis

mp

175 °C (lit.)

SMILES string

[Ga+3].[H]O[H].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/3ClHO4.Ga.H2O/c3*2-1(3,4)5;;/h3*(H,2,3,4,5);;1H2/q;;;+3;/p-3

InChI key

POVMVMOENAMBPV-UHFFFAOYSA-K

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Ox. Sol. 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service