Skip to Content
Merck
All Photos(1)

Documents

410527

Sigma-Aldrich

Gallium(III) sulfide

99.99% trace metals basis

Synonym(s):

Gallium sesquisulfide GaS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ga2S3
CAS Number:
Molecular Weight:
235.64
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

reaction suitability

reagent type: catalyst
core: gallium

density

3.65 g/mL at 25 °C (lit.)

SMILES string

S=[Ga]S[Ga]=S

InChI

1S/2Ga.3S

InChI key

RCKBMGHMPOIFND-UHFFFAOYSA-N

General description

Ga2S3 possesses high energy density, electrochemical study of the compound was studied. Bacterial oxidation of Ga2S3 was investigated.

Application

Gallium(III) sulfide (Ga2S3) may be used in lithium secondary batteries.

Packaging

1 g packaged in glass bottles; 5 g packaged in poly bottles

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Torma
Canadian journal of microbiology, 24(7), 888-891 (1978-07-01)
The bacterial oxidation of naturally occurring gallium-bearing chalcopyrite concentrate and a pure synthetic gallium (III) sulfide has been investigated at pH 1.8 and 35 degree C, using an active culture of Thiobacillus ferrooxidans. This oxidation process may proceed by direct
Gallium (III) sulfide as an active material in lithium secondary batteries
Senoh H, et al.
Journal of Power Sources, 196(13), 5631-5636 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service