Skip to Content
Merck
All Photos(2)

Key Documents

574082

Sigma-Aldrich

Chromium(III) acetylacetonate

99.99% trace metals basis

Synonym(s):

Chromium(III) 2,4-pentanedionate, Cr(acac)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cr(C5H7O2)3
CAS Number:
Molecular Weight:
349.32
Beilstein:
4148971
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

solid

reaction suitability

core: chromium

bp

340 °C (lit.)

mp

210 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Cr](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Cr/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

JWORPXLMBPOPPU-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Chromium(III) acetylacetonate is an acidopentamminechromium (111) salt, which is a crystalline material that can be prepared by treatment of precipitated hydrous chromium(III) oxide with acetylacetone.[1]

Application

Chromium(III) acetylacetonate may be used as an active site in the formation of redox systems for the fabrication of flow batteries.[2] It may also be used as a precursor for chromium nitride based metallic coatings.[3]

Analysis Note

Used to modify the surface properties of solid polyurethanes formed in its presence.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>392.0 °F

Flash Point(C)

> 200 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromium (III) acetylacetonate
Fernelius WC, et al.
Inorganic Syntheses, 5, 130-131 (1957)
Non-aqueous chromium acetylacetonate electrolyte for redox flow batteries
Liu Q, et al.
Electrochemical Communications, 12(11), 1634-1637 (2010)
Plasma assisted metal-organic chemical vapor deposition of hard chromium nitride thin film coatings using chromium (III) acetylacetonate as the precursor
Dasgupta A, et al.
Materials Science & Engineering. A, Structural Materials : Properties, Microstructure and Processing, 374(1-2), 362-368 (2004)
Chiara Allegretti et al.
Molecules (Basel, Switzerland), 25(12) (2020-06-27)
Technical lignins, typically obtained from the biorefining of lignocellulosic raw materials, represent a highly abundant natural aromatic feedstock with high potential in a sustainable economy scenario, especially considering the huge primary production volumes and the inherently renewable nature of this
Zhan'ao Tan et al.
Physical chemistry chemical physics : PCCP, 14(42), 14589-14595 (2012-09-28)
A solution-processed vanadium oxide (s-VO(x)) anode buffer layer on an indium-tin-oxide (ITO) electrode was used instead of PEDOT:PSS for improving the stability and photovoltaic performance of the polymer solar cells (PSCs). The s-VO(x) layer was prepared by spin-coating a vanadyl

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service