Skip to Content
Merck
All Photos(2)

Key Documents

C83902

Sigma-Aldrich

Cobalt(III) acetylacetonate

98%

Synonym(s):

Co(acac)3, Cobalt(III) 2,4-pentanedionate, Cobaltic acetylacetonate, Tris(2,4-pentanedionato)cobalt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€72.20
100 G
€201.00

€72.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€72.20
100 G
€201.00

About This Item

Linear Formula:
Co(C5H7O2)3
CAS Number:
Molecular Weight:
356.26
Beilstein:
9007210
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€72.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder or crystals

reaction suitability

core: cobalt

mp

210-213 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Co](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Co/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

RHCQEPWEBDOALW-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Used for the preparation of a new family of monoporphyrinates which are interesting for their photochemical and magnetic properties and synthetic applications.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I K Adzamli et al.
The Biochemical journal, 211(1), 219-226 (1983-04-01)
Kinetic results are presented for the reaction of reduced [2Fe-2S] ferredoxin from the blue-green alga Spirulina platensis with Co(NH3)6(3+), Co(edta)- and Co(acac)3 as oxidants at pH 8.0 at I0.10 (NaCl). The aim is to compare results obtained with those previously
Lili Zhang et al.
ACS nano, 11(5), 4483-4493 (2017-04-14)
In order to controllably grow single-wall carbon nanotubes (SWCNTs), a better understanding of the growth processes and how they are influenced by external parameters such as catalyst and gaseous environment is required. Here, we present direct evidence of growth termination
Akira Ohashi et al.
Talanta, 146, 789-794 (2015-12-24)
The solubilities of several cobalt(III) chelates (CoL3) with in supercritical carbon dioxide (SC-CO2) were investigated in the presence of fluorine- and trifluoromethyl-substituted phenols (PhOH) using UV-vis spectrophotometry. Solubility enhancement of CoL3 complexes in SC-CO2 was accomplished by adding PhOH. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service