Skip to Content
Merck
All Photos(3)

Key Documents

571326

Sigma-Aldrich

(4S)-(+)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane

96%

Synonym(s):

(S)-(+)-2,2-Dimethyl-1,3-dioxolane-4-ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€310.00
25 G
€404.00

€310.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€310.00
25 G
€404.00

About This Item

Empirical Formula (Hill Notation):
C7H14O3
CAS Number:
Molecular Weight:
146.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€310.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

optical activity

[α]20/D +2°, c = 1% in chloroform

refractive index

n20/D 1.4380 (lit.)

bp

98 °C/15 mmHg (lit.)

density

1.027 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl
ketal

storage temp.

2-8°C

SMILES string

CC1(C)OC[C@H](CCO)O1

InChI

1S/C7H14O3/c1-7(2)9-5-6(10-7)3-4-8/h6,8H,3-5H2,1-2H3/t6-/m0/s1

InChI key

YYEZYENJAMOWHW-LURJTMIESA-N

Application

(4S)-(+)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane may be used in the preparation (prop-2-ynyloxy)ethyl-tethered dioxolanes via O-propargylation.[1] It may also be used as an intermediate in the total synthesis of rugulactone.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.1 °F - closed cup

Flash Point(C)

100.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric total synthesis of rugulactone, an a-pyrone from Cryptocarya rugulosa.
Allais F, et al.
Synthesis, 2010(16), 2787-2793 (2010)
Gold-catalyzed direct cycloketalization of acetonide-tethered alkynes in the presence of water.
Alcaide B, et al.
Tetrahedron, 68(46), 9391-9396 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service