567086
Methyl 3-[(2-methoxy-2-oxoethyl)thio]propionate
95%
Synonym(s):
Methyl 3-[(2-methoxy-2-oxoethyl)thio]propanoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H3CO2CCH2SCH2CH2CO2CH3
CAS Number:
Molecular Weight:
192.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
refractive index
n20/D 1.4750 (lit.)
bp
258-259 °C (lit.)
bulk density
1.154 g/mL
functional group
ester
thioether
SMILES string
COC(=O)CCSCC(=O)OC
InChI
1S/C7H12O4S/c1-10-6(8)3-4-12-5-7(9)11-2/h3-5H2,1-2H3
InChI key
XNDKLLFGXIEGKL-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
129.9 °F - closed cup
Flash Point(C)
54.4 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service