Skip to Content
Merck
All Photos(2)

Documents

565733

Sigma-Aldrich

Poly(ethylene oxide)

average MN 10,000, 4-arm, amine terminated, carboxyl reactive

Synonym(s):

Polyethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene oxide), 4-arm, amine terminated, average Mn 10,000

form

solid

mol wt

average Mn 10,000

reaction suitability

reactivity: carboxyl reactive

mp

50-54 °C (lit.)

Mw/Mn

<1.2

polymer architecture

shape: 4-arm
functionality: homofunctional

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service