Skip to Content
Merck
All Photos(4)

Key Documents

372781

Sigma-Aldrich

Poly(ethylene oxide)

average MV 1,000,000 (nominal), powder, hydroxyl, BHT as inhibitor

Synonym(s):

Polyethylene oxide, PEO

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€67.00
250 G
€200.00
500 G
€366.00

€67.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€67.00
250 G
€200.00
500 G
€366.00

About This Item

Linear Formula:
(-CH2CH2O-)n
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

€67.00


Check Cart for Availability

Request a Bulk Order

Product Name

Poly(ethylene oxide), average Mv ~1,000,000 (nominal), powder

form

powder

Quality Level

mol wt

average Mv ~1,000,000 (nominal)

contains

200-500 ppm BHT as inhibitor

viscosity

400-800 cP, 2 % in H2O(25 °C, Brookfield)(lit.)

functional group

hydroxyl

SMILES string

[H]OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene oxide) (PEO) is a crystalline, thermoplastic, water-soluble polymer that is commercially available.[1]

Application

PEO (Mv ~ 1,000,000, powder) can be used in the fabrication of hollow TiO2 fibers, which can be used for a variety of applications such as drug delivery, catalysts, sensors, and electric devices.[1] It can also be used as an electrolyte for the formation of dye-sensitized solar cells (DSSCs).[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-state dye-sensitized hierarchically structured ZnO solar cells
Deng J, et al.
Electrochimica Acta, 56(11), 4176-4180 (2011)
Fabrication of hollow TiO2 fibers templated by electrospun aqueous poly (ethylene oxide)(PEO) solution
Nagamine S, et al.
Chemistry Letters (Jpn), 38(3), 258-259 (2009)
Yi Huang et al.
Polymers, 12(5) (2020-05-21)
Controlling the rheological property of suspensions consisting of colloidal particles and polymers is necessary in industry. Especially, gels induced by shear (shake-gel) are interesting phenomena in rheological field. To gain insight into the shake-gel phenomena of the aqueous suspensions of
Ievgenii Liashenko et al.
Nature communications, 11(1), 753-753 (2020-02-08)
Additive manufacturing technologies based on layer-by-layer deposition of material ejected from a nozzle provide unmatched versatility but are limited in terms of printing speed and resolution. Electrohydrodynamic jetting uniquely allows generating submicrometer jets that can reach speeds above 1 m s-1, but
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced

Articles

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service