Skip to Content
Merck
All Photos(2)

Key Documents

556912

Sigma-Aldrich

Isooctyl 3-mercaptopropionate

≥99%

Synonym(s):

Isooctyl β-mercaptopropionate, Isooctyl mercaptopropionate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€71.90

€71.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€71.90

About This Item

Linear Formula:
HS-CH2CH2COOC8H17
CAS Number:
Molecular Weight:
218.36
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€71.90


Check Cart for Availability

Request a Bulk Order

Assay

≥99%

refractive index

n20/D 1.458 (lit.)

bp

109-112 °C/2 mmHg (lit.)

density

0.950 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCCOC(=O)CCS

InChI

1S/C11H22O2S/c1-10(2)6-4-3-5-8-13-11(12)7-9-14/h10,14H,3-9H2,1-2H3

InChI key

ZHUWXKIPGGZNJW-UHFFFAOYSA-N

Application

Polymerization modifier: crosslinking agent, chain transfer agent

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service