Skip to Content
Merck
All Photos(1)

Key Documents

284394

Sigma-Aldrich

N,N-Dimethylglycine ethyl ester

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€128.00

€128.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€128.00

About This Item

Linear Formula:
(CH3)2NCH2COOC2H5
CAS Number:
Molecular Weight:
131.17
Beilstein:
1750085
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€128.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.413 (lit.)

bp

150-151 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCOC(=O)CN(C)C

InChI

1S/C6H13NO2/c1-4-9-6(8)5-7(2)3/h4-5H2,1-3H3

InChI key

BGCNBOFPABQGNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tzi-Yi Wu et al.
International journal of molecular sciences, 12(12), 8750-8772 (2012-01-25)
This work includes specific basic characterization of synthesized glycine-based Ionic Liquid (IL) [QuatGly-OEt][EtOSO(3)] by NMR, elementary analysis and water content. Thermophysical properties such as density, ρ, viscosity, η, refractive index, n, and conductivity, κ, for the binary mixture of [QuatGly-OEt][EtOSO(3)]

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service