Skip to Content
Merck
All Photos(3)

Key Documents

555975

Sigma-Aldrich

3,5-Difluorobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3SO2Cl
CAS Number:
Molecular Weight:
212.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

SMILES string

Fc1cc(F)cc(c1)S(Cl)(=O)=O

InChI

1S/C6H3ClF2O2S/c7-12(10,11)6-2-4(8)1-5(9)3-6/h1-3H

InChI key

IIQKIICAOXAXEJ-UHFFFAOYSA-N

Application

3,5-Difluorobenzenesulfonyl chloride may be used to synthesize the following:
  • N,N-diethyl-3,5-difluorobenzene sulfonamide
  • N-phenyl-3,5-difluorobenzene sulfonamide
It can be used for the regioselective monosulfonation of methyl-α-D-glucopyranoside.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Utilization of N, N-diethyl-3, 5-difluorobenzene sulfonamide to prepare functionalized poly (arylene ether) s.
Selhorst R and Fossum E.
Polymer, 54(2), 530-535 (2013)
"Chemo-and regioselective monosulfonylation of nonprotected carbohydrates catalyzed by organotin dichloride under mild conditions"
Muramatsu W
The Journal of Organic Chemistry, 77(18), 8083-8091 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service