Skip to Content
Merck
All Photos(1)

Key Documents

551473

Sigma-Aldrich

QuadraPure® AK

50-90 mesh, extent of labeling: 2.0-3.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

2,4-Butanedione resin, Activated ketone, polymer-bound

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€262.00

€262.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€262.00

About This Item

UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€262.00


Check Cart for Availability

Request a Bulk Order

product line

QuadraPure®

Quality Level

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: nucleophile reactive

extent of labeling

2.0-3.0 mmol/g loading

particle size

50-90 mesh

Application

QuadraPure® AK is a resin which is generally used as a support for heterogeneous catalysis.[1] It can be used as a hydroxylamine scavenger.[2]
Scavenger for Hydrazines and Primary Amines

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu, Z. et al.
Tetrahedron Letters, 41, 8963-8963 (2000)
Quadrapure-supported palladium nanocatalysts for microwave-promoted Suzuki cross-coupling reaction under aerobic condition
Liew KH, et al.
TheScientificWorldJournal, 2014 (2014)
Flow synthesis of tricyclic spiropiperidines as building blocks for the histrionicotoxin family of alkaloids
Brasholz M, et al.
Tetrahedron, 66(33), 6445-6449 (2010)
A Polymer-Bound 1,3-Diketone: A Highly Efficient Scavenger for Hydrazines, and Primary Amines
Uwe Schon, Josef Messinger, Nuria Merayo, Grzegorz Juszkiewicz, Andreas Kirschning
Synlett, 7, 983-983 (2003)
QuadraPure Cartridges for Removal of Trace Metal from Reaction Mixtures in Flow
A. Hinchcliffe, et al.
Organic Process Research & Development, 11, 477-481 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service