Skip to Content
Merck
All Photos(1)

Key Documents

549878

Sigma-Aldrich

4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O4S
CAS Number:
Molecular Weight:
218.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

129-133 °C (lit.)

SMILES string

COc1cc(OC)nc(n1)S(C)(=O)=O

InChI

1S/C7H10N2O4S/c1-12-5-4-6(13-2)9-7(8-5)14(3,10)11/h4H,1-3H3

InChI key

ITDVJJVNAASTRS-UHFFFAOYSA-N

General description

4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine can be prepared by the oxidation of 2-methylthio-4,6-dimethoxypyrimidine.

Application

4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine may be used in the microwave assisted preparation of 4,6-dimethoxy-N-methylpyrimidin-2-amine by reacting with methylamine. It may also be used to prepare 2′-pyrimidinecarbonylsulfonanilide derivatives with potent herbicidal activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tunable push-pull interactions in 5-nitrosopyrimidines.
Procha'zkova' E, et al.
The Journal of Organic Chemistry, 81(9), 3780-3789 (2016)
Synthesis and herbicidal activity of sulfonanilides having a pyrimidinyl-containing group at the 2'-position.
Yoshimura T, et al.
Journal of Pesticide Science, 36(11), 212-220 (2011)
A Facile Synthesis of 4, 6-Dimethoxy-2-methylsulfonylpyrimidine.
Xu D, et al.
Asian Journal of Chemistry, 26(1), 313-313 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service