Skip to Content
Merck
All Photos(1)

Key Documents

549339

Sigma-Aldrich

Decyl chloroformate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€95.50

€95.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€95.50

About This Item

Linear Formula:
ClCO2(CH2)9CH3
CAS Number:
Molecular Weight:
220.74
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€95.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.438 (lit.)

bp

226-227 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

CCCCCCCCCCOC(Cl)=O

InChI

1S/C11H21ClO2/c1-2-3-4-5-6-7-8-9-10-14-11(12)13/h2-10H2,1H3

InChI key

AZZCHVHSWUYCQA-UHFFFAOYSA-N

General description

Decyl chloroformate is an n-alkyl chloroformate derivative that can be prepared by reacting decyl alcohol with triphosgene.[1]

Application

Decyl chloroformate may be used in the preparation of (S)-(+)-N-decoxycarbonyl-leucine, a chiral surfactant.[1] It may also be used to synthesize decyl chloride via decarboxylation in the presence of dimethylformamide.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbamate chiral surfactants for capillary electrophoresis.
Ding W and Fritz JS.
Journal of Chromatography A, 831(2), 311-320 (1999)
Dimethylformamide-catalyzed decarboxylation of alkyl chloroformates. A synthesis of primary alkyl chlorides.
Richter R and Tucker B.
The Journal of Organic Chemistry, 48(15), 2625-2627 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service