Skip to Content
Merck
All Photos(2)

Key Documents

237906

Sigma-Aldrich

Phenyl chloroformate

97%

Synonym(s):

Chloroformic acid phenyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€84.50
100 G
€147.00

€84.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€84.50
100 G
€147.00

About This Item

Linear Formula:
ClCOOC6H5
CAS Number:
Molecular Weight:
156.57
Beilstein:
606778
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€84.50


Check Cart for Availability

Request a Bulk Order

vapor density

1 (vs air)

Quality Level

vapor pressure

1.22 psi ( 20 °C)
1.67 psi ( 55 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

74-75 °C/13 mmHg (lit.)

density

1.248 g/mL at 25 °C (lit.)

functional group

chloro
phenoxy

storage temp.

2-8°C

SMILES string

ClC(=O)Oc1ccccc1

InChI

1S/C7H5ClO2/c8-7(9)10-6-4-2-1-3-5-6/h1-5H

InChI key

AHWALFGBDFAJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics of spontaneous hydrolysis of phenyl chloroformate has been studied in water-ethylene glycol, cationic, zwitterionic, nonionic and anionic micellar solutions[1].

Application

Phenyl chloroformate has been used in the synthesis of poly(2-(phenoxycarbonyloxy)ethylmethacrylate)[2] and phenyl-(4-vinylphenyl)carbonate[3].

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and ligand recognition of paracetamol selective polymers: semi-covalent versus non-covalent molecular imprinting.
Rosengren-Holmberg JP, et al.
Organic & Biomolecular Chemistry, 7(15), 3148-3155 (2009)
Amalia Rodríguez et al.
Langmuir : the ACS journal of surfaces and colloids, 20(23), 9945-9952 (2004-11-03)
The spontaneous hydrolysis of phenyl chloroformate was studied in water-ethylene glycol, EG, cationic, zwitterionic, nonionic, and anionic micellar solutions, the surfactants being tetradecyltrimethylammonium bromide, tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, tricosaoxyethylene glycol ether, and sodium dodecyl sulfate. The dependence of the observed rate constant on
K Anzai et al.
Nucleic acids symposium series, (15)(15), 49-52 (1984-01-01)
Reaction of 2',3'-O-isopropylideneadenosine with p-toluyl chloride in a mixture of methylene chloride and triethylamine afforded 2',3'-O-isopropylidene-N,N,5'-O-tri-p-toluyladenosine (8)(30%), 8,5'-O-cycloadenosine derivatives 9 (34%) and 10 (11%), and a cyanoimidazole nucleoside 11 (12%).
B Boettcher et al.
Analytical biochemistry, 138(2), 449-450 (1984-05-01)
An improved synthesis of L-2- oxothiazolidine -4-carboxylic acid is described. The new procedure, which leads to excellent yields of product, does not require the use of phosgene. The new method is thus less hazardous than the original one, and is
Luis García-Río et al.
Langmuir : the ACS journal of surfaces and colloids, 21(17), 7672-7679 (2005-08-11)
A study was carried out on the solvolysis of substituted phenyl chloroformates in AOT/isooctane/water microemulsions. (AOT is the sodium salt of bis(2-ethyhexyl)sulfosuccinate.) The results obtained have been interpreted by taking into account the distribution of the chloroformates between the continuous

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service