Skip to Content
Merck
All Photos(1)

Key Documents

549169

Sigma-Aldrich

5-Amino-3-(4-methylphenyl)-1-phenylpyrazole

97%

Synonym(s):

3-(4-Methylphenyl)-1-phenyl-1H-pyrazol-5-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3
CAS Number:
Molecular Weight:
249.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

173-176 °C (lit.)

SMILES string

Cc1ccc(cc1)-c2cc(N)n(n2)-c3ccccc3

InChI

1S/C16H15N3/c1-12-7-9-13(10-8-12)15-11-16(17)19(18-15)14-5-3-2-4-6-14/h2-11H,17H2,1H3

InChI key

WMJZITXAXMNGCH-UHFFFAOYSA-N

General description

5-Amino-3-(4-methylphenyl)-1-phenylpyrazole (3-(4-methylphenyl)-1-phenyl-1H-pyrazol-5-amine) belongs to a family of 5-aminopyrazole derivatives, which are bioactive agents. They find a wide range of applications in the pharmaceutical and agrochemical industries. It can be prepared by refluxing a mixture of p-methyl-benzoylacetonitrile and phenylhydrazine in ethanol.

Application

5-Amino-3-(4-methylphenyl)-1-phenylpyrazole may be used in the preparation of N′-[4-formyl-3-(4-methylphenyl)-1-phenyl-1H-pyrazol-5-yl]-N,N-dimethylimidoformamide, an intermediate for preparing 5-amino-3-(4-methylphenyl)-1-phenyl-1H-pyrazole-4-carbaldehyde.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Friedlander condensation of 5-aminopyrazole-4-carbaldehydes with reactive a-methylene ketones: Synthesis of pyrazolo [3,4-b] pyridines.
Jachak MN, et al.
Journal of Heterocyclic Chemistry, 42(7), 1311-1319 (2005)
Approaches towards the synthesis of 5-aminopyrazoles.
Aggarwal R, et al.
Beilstein Journal of Organic Chemistry, 7(1), 179-197 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service