Skip to Content
Merck
All Photos(3)

Key Documents

C28006

Sigma-Aldrich

(4-Chlorophenyl)acetonitrile

96%

Synonym(s):

4-Chlorobenzyl cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€43.50

€43.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€43.50

About This Item

Linear Formula:
ClC6H4CH2CN
CAS Number:
Molecular Weight:
151.59
Beilstein:
971171
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€43.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid (or Semi-Solid or Liquid)

bp

265-267 °C (lit.)

mp

25-28 °C (lit.)

density

1.19 g/mL at 20 °C (lit.)

SMILES string

Clc1ccc(CC#N)cc1

InChI

1S/C8H6ClN/c9-8-3-1-7(2-4-8)5-6-10/h1-4H,5H2

InChI key

IVYMIRMKXZAHRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

271.4 °F - closed cup

Flash Point(C)

133 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service