Skip to Content
Merck
All Photos(1)

Key Documents

542695

Sigma-Aldrich

5-Chlorothiophene-2-sulfonamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4ClNO2S2
CAS Number:
Molecular Weight:
197.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

113-117 °C (lit.)

SMILES string

NS(=O)(=O)c1ccc(Cl)s1

InChI

1S/C4H4ClNO2S2/c5-3-1-2-4(9-3)10(6,7)8/h1-2H,(H2,6,7,8)

InChI key

RKLQLYBJAZBSEU-UHFFFAOYSA-N

General description

5-Chlorothiophene-2-sulfonamide is an aromatic sulfonamide. It undergoes Rh-catalyzed aerobic N-alkylation with benzyl alcohol to yield the corresponding N-alkylated sulfonamide.[1]

Application

5-Chlorothiophene-2-sulfonamide may be used in the synthesis of non-benzofused bicyclo[4.2.1]nonanes.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tim Sparey et al.
Bioorganic & medicinal chemistry letters, 18(1), 375-379 (2007-12-07)
Bridgehead substituted derivatives of bicyclo[4.2.1]nonanes were synthesized and shown to be potent inhibitors of gamma-secretase. Two related series were synthesized to explore the SARs. More potent compounds were found in the non-benzofused series compared with the benzofused series. One compound
Rhodium-catalyzed aerobic N-alkylation of sulfonamides with alcohols.
Feng SL, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 22(9), 1021-1024 (2011)
Zuhal Alım et al.
Pharmacological reports : PR, 72(6), 1738-1748 (2020-08-05)
Thiophene(s) are an important group in therapeutic applications, and sulfonamides are the most important class of carbonic anhydrase (CA) inhibitors. In this study, inhibition effects of some thiophene-based sulfonamides on human erythrocytes carbonic anhydrase I and II isoenzymes (hCA-I and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service