Skip to Content
Merck
All Photos(1)

Key Documents

540706

Sigma-Aldrich

Tris(acetonitrile)tricarbonyltungsten(0)

Synonym(s):

Tris(acetonitrile)tungsten(0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
W(NCCH3)3(CO)3
CAS Number:
Molecular Weight:
391.03
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: tungsten
reagent type: catalyst

mp

>300 °C (lit.)

SMILES string

[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].CC#N.CC#N.CC#N

InChI

1S/3C2H3N.3CO.W/c3*1-2-3;3*1-2;/h3*1H3;;;;

InChI key

MHVAYSUFFMSBKA-UHFFFAOYSA-N

Application

Catalyst for allylic substitutions

Reactant for:
  • Mixed disilyl-substituted cyclopentadiene derivatives
  • Synthesis of catalytically active ansa compounds with cycloalkyl moieties
  • Stepwise assembly of mixed metal dinuclear carbonyl complexes
  • Cycloaddition of M(CO)3-coordinated-cyanoazepine with alkynes

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service