Skip to Content
Merck
All Photos(3)

Key Documents

919349

Sigma-Aldrich

NaDT

Synonym(s):

Sodium decatungstate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€114.00
1 G
€370.00

€114.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€114.00
1 G
€370.00

About This Item

Empirical Formula (Hill Notation):
Na4O32W10
CAS Number:
Molecular Weight:
2442.34
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

€114.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

Application

NaDT is a decatungstate photocatalyst with demonstrated uses in the trifluoromethylation of aliphatic C-H bonds as wells the selective C-H fluorination of unprotected peptides.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick J Sarver et al.
Nature chemistry, 12(5), 459-467 (2020-03-24)
The introduction of a trifluoromethyl (CF3) group can dramatically improve a compound's biological properties. Despite the well-established importance of trifluoromethylated compounds, general methods for the trifluoromethylation of alkyl C-H bonds remain elusive. Here we report the development of a dual-catalytic
Zheliang Yuan et al.
Angewandte Chemie (International ed. in English), 57(39), 12733-12736 (2018-08-08)
Peptides are often ideal ligands for diagnostic molecular imaging due to their ease of synthesis and tuneable targeting properties. However, labelling unmodified peptides with 18 F for positron emission tomography (PET) imaging presents a number of challenges. Here we show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service