Skip to Content
Merck
All Photos(1)

Key Documents

540048

Sigma-Aldrich

(R)-(+)-Propylene oxide

99%

Synonym(s):

(R)-(+)-1,2-Epoxypropane, (R)-(+)-Methyloxirane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€162.00
25 G
€266.00

€162.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€162.00
25 G
€266.00

About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
Beilstein:
79764
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€162.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

optical activity

[α]20/D +14°, neat

refractive index

n20/D 1.366 (lit.)

bp

33-34 °C (lit.)

density

0.829 g/mL at 20 °C (lit.)

functional group

ether

SMILES string

C[C@@H]1CO1

InChI

1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3/t3-/m1/s1

InChI key

GOOHAUXETOMSMM-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(+)-Propylene oxide is commonly employed as a chiral building block or intermediate in the synthesis of chiral compounds. For example, it can be used to synthesize cyclic propylene carbonate by the reaction with CO2 in the presence of a chiral catalyst. It is also used as a starting material for the preparation of magnetic resonance (MR) contrast agents that can be used for in vivo detection of β-galactosidase.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F - Pensky-Martens closed cup

Flash Point(C)

-40 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steffi Daniel et al.
Journal of molecular medicine (Berlin, Germany), 98(11), 1639-1656 (2020-09-24)
Fibulin-3 (F3) is an extracellular matrix glycoprotein found in basement membranes across the body. An autosomal dominant R345W mutation in F3 causes a macular dystrophy resembling dry age-related macular degeneration (AMD), whereas genetic removal of wild-type (WT) F3 protects mice
Nagore Puente et al.
Bio-protocol, 9(2), e3145-e3145 (2019-01-20)
Activation of type 1 cannabinoid (CB1) receptors by endogenous, exogenous (cannabis derivatives) or synthetic cannabinoids (i.e., CP 55.940, Win-2) has a wide variety of behavioral effects due to the presence of CB1 receptors in the brain. In situ hybridization and
Amanda S Hakemian et al.
Biochemistry, 47(26), 6793-6801 (2008-06-11)
Particulate methane monooxygenase (pMMO) is a membrane-bound metalloenzyme that oxidizes methane to methanol in methanotrophic bacteria. The nature of the pMMO active site and the overall metal content are controversial, with spectroscopic and crystallographic data suggesting the presence of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service