Skip to Content
Merck
All Photos(1)

Key Documents

539260

Sigma-Aldrich

3-Glycidoxypropyldimethylethoxysilane

97%

Synonym(s):

3-[(Glycidyloxy)propyl]dimethylethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22O3Si
CAS Number:
Molecular Weight:
218.37
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

100 °C/3 mmHg (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](C)(C)CCCOCC1CO1

InChI

1S/C10H22O3Si/c1-4-13-14(2,3)7-5-6-11-8-10-9-12-10/h10H,4-9H2,1-3H3

InChI key

HHBOIIOOTUCYQD-UHFFFAOYSA-N

Application

3-Glycidoxypropyldimethylethoxysilane was used to functionalize glass substrates to study fluorescence enhancement performance of fabricated photonic crystal sensor. The product could be used for the silanization of hydroxlated Si(100) wafers, and optical chips. Glycidoxypropyldimethylethoxysilane, along with several silanes, was used to form bonded phases for supercritical fluid chromatography (SFC).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

189.0 °F - closed cup

Flash Point(C)

87.22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hartman interferometer: versatile integrated optic sensor for label-free, real-time quantification of nucleic acids, proteins, and pathogens.
Schneider BH, et al.
Clinical Chemistry, 43(9), 1757-1763 (1997)
Richard J Sheridan et al.
Macromolecules, 50(17), 6668-6678 (2017-10-04)
We present a method for the direct measurement of the relative energy of interaction between a solvated polymer and a solid interface. By tethering linear chains covalently to the surface, we ensured the idealized and constant configuration of polymer molecules
Multicolor fluorescence enhancement from a photonics crystal surface.
Pokhriyal A, et al.
Applied Physics Letters, 97(2), 121108-121108 (2010)
STM and HREELS investigation of gas phase silanization on hydroxylated Si (100).
Fan C and Lopinski GP
Surface Science, 604(11), 996-1001 (2010)
Silica surface interactions of diol?bonded phases in packed capillary column supercritical fluid chromatography
Shen Y and Lee ML
J. Microcol. Sep., 8(6), 413-420 (1996)

Articles

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service