Skip to Content
Merck
All Photos(1)

Documents

53586

Sigma-Aldrich

(S)-2-Methyl-1,4-butanediol

≥97.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O2
CAS Number:
Molecular Weight:
104.15
Beilstein:
2343656
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (sum of enantiomers, GC)

refractive index

n20/D 1.449

density

0.992 g/mL at 20 °C (lit.)

SMILES string

C[C@H](CO)CCO

InChI

1S/C5H12O2/c1-5(4-7)2-3-6/h5-7H,2-4H2,1H3/t5-/m0/s1

InChI key

MWCBGWLCXSUTHK-YFKPBYRVSA-N

Application

Used in the synthesis of sex pheromones of pine sawflies.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei-Qiang Huang et al.
The Journal of organic chemistry, 69(11), 3964-3967 (2004-05-22)
(2S,3S,7S)-3,7-Dimethylpentadecan-2-yl acetate (2) and its propionate analogue (3) are the main sex pheromones of all Neodiprion species and Diprion similes, respectively. Starting from (S)-malic acid and employing a highly chemo-, regio-, and stereoselective tandem ester reduction-epoxide formation-reductive epoxide-opening reaction protocol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service