Skip to Content
Merck
All Photos(2)

Key Documents

327220

Sigma-Aldrich

2,2,4-Trimethyl-1,3-pentanediol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€51.00

€51.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€51.00

About This Item

Linear Formula:
(CH3)2CHCH(OH)C(CH3)2CH2OH
CAS Number:
Molecular Weight:
146.23
Beilstein:
1698098
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€51.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

bp

232 °C (lit.)

mp

50-53 °C (lit.)

SMILES string

CC(C)C(O)C(C)(C)CO

InChI

1S/C8H18O2/c1-6(2)7(10)8(3,4)5-9/h6-7,9-10H,5H2,1-4H3

InChI key

JCTXKRPTIMZBJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrian Brenz et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(2), 307-319 (2016-11-29)
Polyesters (PESs) are gaining more importance on the food contact material (FCM) market and the variety of properties and applications is expected to be wide. In order to acquire the desired properties manufacturers can combine several FCM-approved polyvalent carboxylic acids
Akiko Fujita et al.
PloS one, 15(2), e0229269-e0229269 (2020-02-23)
Psychiatric disorders including depression and anxiety comprise a broad range of conditions with different symptoms. We have developed a mouse model of depression/anxiety in mice deficient in the St3gal4 gene. In this study, we performed a comparative analysis of urinary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service