Skip to Content
Merck
All Photos(3)

Key Documents

535508

Sigma-Aldrich

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€22.90
25 G
€31.90

€22.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€22.90
25 G
€31.90

About This Item

Empirical Formula (Hill Notation):
C9H12ClNO · HCl
CAS Number:
Molecular Weight:
222.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€22.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

128-131 °C (lit.)

functional group

chloro

SMILES string

Cl[H].COc1c(C)cnc(CCl)c1C

InChI

1S/C9H12ClNO.ClH/c1-6-5-11-8(4-10)7(2)9(6)12-3;/h5H,4H2,1-3H3;1H

InChI key

LCJDHJOUOJSJGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Li et al.
Toxicology in vitro : an international journal published in association with BIBRA, 41, 159-167 (2017-02-22)
The in vitro EpiSkin™ test method was validated in 2007 by the European Union Reference Laboratory for alternatives to animal testing (EURL ECVAM) as a full replacement method for the Draize acute skin irritation test and adopted in the OECD

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service