Skip to Content
Merck
All Photos(2)

Key Documents

529559

Sigma-Aldrich

Cerium(III) acetate hydrate

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€76.80

€76.80


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
€76.80

About This Item

Linear Formula:
Ce(CH3CO2)3 · xH2O
CAS Number:
Molecular Weight:
317.25 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€76.80


Estimated to ship on12 April 2025


Request a Bulk Order

Quality Level

Assay

99.99% trace metals basis

form

powder or crystals

reaction suitability

core: cerium
reagent type: catalyst

SMILES string

O.CC(=O)O[Ce](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.Ce.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3

InChI key

AERUOEZHIAYQQL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service