Skip to Content
Merck
All Photos(2)

Key Documents

526134

Sigma-Aldrich

tert-Butyldimethylsilyl glycidyl ether

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€314.00

€314.00


Estimated to ship on10 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
€314.00

About This Item

Empirical Formula (Hill Notation):
C9H20O2Si
CAS Number:
Molecular Weight:
188.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€314.00


Estimated to ship on10 April 2025


Request a Bulk Order

Assay

98%

refractive index

n20/D 1.531 (lit.)

bp

195-197 °C (lit.)

density

0.901 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CC(C)(C)[Si](C)(C)OCC1CO1

InChI

1S/C9H20O2Si/c1-9(2,3)12(4,5)11-7-8-6-10-8/h8H,6-7H2,1-5H3

InChI key

YANSSVVGZPNSKD-UHFFFAOYSA-N

General description

tert-Butyldimethylsilyl glycidyl ether [tert-butyl-dimethyl-(oxiran-2-ylmethoxy)silane] can be synthesized from glycidol, via hydrolytic kinetic resolution (HKR) in the presence of R-(salen)Co complex and water.[1]

Application

tert-Butyldimethylsilyl glycidyl ether may be used to synthesize (R/S)-1-benzylamino-3-(tert-butyldimethylsilyloxy)propan-2-ol and (2R/2S)-1-(tert-butyldimethylsilyloxy)-3-{[(4S)-2,2-dimethyl-1,3-dioxolan-4-ylmethyl]amino}propan-2-ol.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical access to highly enantioenriched C-3 building blocks via hydrolytic kinetic resolution.
Furrow ME, et al.
The Journal of Organic Chemistry, 63(20), 6776-6777 (1998)
Synthesis of Glycidol-and Sugar-Derived Bicyclic ?-and ?/d-Amino Acids for Peptidomimetic Design.
Danieli E, et al.
European Journal of Organic Chemistry, 2005(20), 4372-4381 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service