Skip to Content
Merck
All Photos(1)

Key Documents

52586

Sigma-Aldrich

1,1,1,3,3,3-Hexakis(dimethylamino)diphosphazenium tetrafluoroborate

≥98.0% (T)

Synonym(s):

Bis[tris(dimethylamino)phosphoranylidene]ammonium tetrafluoroborate, Phosphazenium tetrafluoroborate P2-BF4

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€188.00

€188.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€188.00

About This Item

Linear Formula:
[[(CH3)2N]3P=]2N(BF4)
CAS Number:
Molecular Weight:
427.21
Beilstein:
5470923
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€188.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (T)

form

solid

mp

≥260 °C (lit.)

functional group

amine

SMILES string

F[B-](F)(F)F.CN(C)P(=[N+]=P(N(C)C)(N(C)C)N(C)C)(N(C)C)N(C)C

InChI

1S/C12H36N7P2.BF4/c1-14(2)20(15(3)4,16(5)6)13-21(17(7)8,18(9)10)19(11)12;2-1(3,4)5/h1-12H3;/q+1;-1

InChI key

DPIDWCQJGWFENO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Phase transfer catalyst directing enolate alkylation towards O-substitution[1]; Starting material for preparing the corresponding fluoride, an extremely reactive naked fluoride, with KF[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.V. Dehmlow et al.
Chemische Berichte, 126, 2765-2765 (1993)
R. Schwesinger et al
Angewandte Chemie (International Edition in English), 103, 1376-1376 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service