Skip to Content
Merck
All Photos(1)

Documents

139947

Sigma-Aldrich

2,3-Dibromopropionic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2BrCHBrCOOH
CAS Number:
Molecular Weight:
231.87
Beilstein:
1721428
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

160 °C/20 mmHg (lit.)

mp

64-66 °C (lit.)

SMILES string

OC(=O)C(Br)CBr

InChI

1S/C3H4Br2O2/c4-1-2(5)3(6)7/h2H,1H2,(H,6,7)

InChI key

ZMYAKSMZTVWUJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2,3-Dibromopropionic acid was used in chemical shift imaging during analysis of multiple samples by multiplex sample NMR methodology. It was used as surrogate standard during extraction and determination of haloacetic acid in drinking water.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occurrence and Determination of Haloacetic Acids in Metro Manila Drinking Water.
Rodriguez IB and Espino MPB.
Science Diliman, 21(2), 35-41 (2010)
Carles Planas et al.
Analytical and bioanalytical chemistry, 411(17), 3905-3917 (2019-06-04)
A fast, simple, selective, and sensitive method for the analysis of 11 haloacetic acids (HAAs) in chlorine-treated water has been developed. The method is based on liquid chromatography-electrospray ionization-triple quadrupole tandem mass spectrometry (LC/ESI-QqQ-MS/MS) with direct injection of the aqueous
T Hou et al.
Analytical chemistry, 73(11), 2541-2546 (2001-06-14)
Two improved approaches for the rapid analysis of multiple samples using multiplex sample NMR are described. In the first approach, frequency-selective 90 degrees radio frequency pulses and large pulsed field gradients are applied to excite and detect multiple samples in
Tarek Manasfi et al.
International journal of hygiene and environmental health, 220(3), 583-590 (2017-02-16)
An undesirable consequence of disinfection is the formation of chemical contaminants known as disinfection byproducts (DBPs). Chronic exposure to DBPs has been linked to adverse health effects. The occurrence of DBPs in chlorinated pools filled with seawater (such as thalassotherapy
Tarek Manasfi et al.
International journal of hygiene and environmental health, 222(1), 1-8 (2018-07-23)
Chlorination of seawater is one of the most effective technologies for industrial biofouling control. However, chlorination leads to the formation of halogenated chlorination byproducts (CBPs) associated with potential risks to environmental and human health. The present study investigated the occurrence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service