Skip to Content
Merck
All Photos(1)

Key Documents

522449

Sigma-Aldrich

α,4-Dimethylphenylacetic acid

97%

Synonym(s):

2-(4-Methylphenyl)propanoic acid, p-Methylhydratropic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(CH3)CO2H
CAS Number:
Molecular Weight:
164.20
Beilstein:
2251315
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

37-42 °C (lit.)

SMILES string

CC(C(O)=O)c1ccc(C)cc1

InChI

1S/C10H12O2/c1-7-3-5-9(6-4-7)8(2)10(11)12/h3-6,8H,1-2H3,(H,11,12)

InChI key

KDYOFXPLHVSIHS-UHFFFAOYSA-N

Gene Information

human ... IL8RA(3577)

Looking for similar products? Visit Product Comparison Guide

General description

α,4-Dimethylphenylacetic acid is also referred to as 2-(4-methylphenyl)propanoic acid. It is an alkaline decomposition product of azalomycin-B, a macrolide antibiotic produced by Streptomyces hygroscopicus var. azalomyceticus. The reaction of pinonic acid with bromine in water affords 2-(4-methylphenyl)propanoic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical Studies on Azalomycins. III. Alkaline Degradation of Azalomycin-B.
Tukahashi S and 0kki E.
Chemical & Pharmaceutical Bulletin, 15(11), 1726-1732 (1967)
Pinonic Acid-a Promising Chemical Raw Material.
Hedrick G and Lawrence R.
Industrial & Engineering Chemistry Fundamentals, 52(10), 853-856 (1960)
T Matsumoto et al.
Biological & pharmaceutical bulletin, 17(11), 1441-1445 (1994-11-01)
4-Isopropenyltoluene (8) was administered orally to rabbits and the following four optically active metabolites, 2-(p-tolyl)propanoic acid (10), p-(1-carboxyethyl)benzoic acid (11), 2-hydroxy-2-(p-tolyl)propanoic acid (12), and 2-(p-tolyl)-1,2-propanediol (13) were isolated from urine in addition to an optically inactive metabolite, 4-isopropenylbenzoic acid (9).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service