Skip to Content
Merck
All Photos(1)

Key Documents

374547

Sigma-Aldrich

3-Methoxypropionic acid

96%

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€190.00

€190.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€190.00

About This Item

Linear Formula:
CH3OCH2CH2COOH
CAS Number:
Molecular Weight:
104.10
Beilstein:
1743053
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€190.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.420 (lit.)

bp

116 °C/9 mmHg (lit.)

density

1.108 g/mL at 25 °C (lit.)

functional group

carboxylic acid
ether

SMILES string

COCCC(O)=O

InChI

1S/C4H8O3/c1-7-3-2-4(5)6/h2-3H2,1H3,(H,5,6)

InChI key

YSIKHBWUBSFBRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methoxypropionic acid is an alkoxy acid. The base-catalyzed methanolysis of 1-nitroso-5,6-dihydrouracil, potent liver carcinogen, is reported to yield 3-methoxypropionic acid.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Mirvish et al.
Journal of the National Cancer Institute, 74(5), 1105-1110 (1985-05-01)
Base-catalyzed hydrolysis of the potent liver carcinogen 1-nitroso-5,6-dihydrouracil [(NDHU) CAS: 16813-36-8] afforded 3-hydroxypropionic acid and acrylic acid in 72 and 6% yield, respectively. The base-catalyzed methanolysis of NDHU gave quantitative yields of 3-methoxypropionic acid and methyl carbamate. These results indicate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service