Skip to Content
Merck
All Photos(1)

Key Documents

522120

Sigma-Aldrich

Dimethyl sulfoxide-d6

99.9 atom % D

Synonym(s):

(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein:
1237248
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

vapor pressure

0.42 mmHg ( 20 °C)

isotopic purity

99.9 atom % D

Assay

99% (CP)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pedro Aqueveque et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 70(3-4), 97-102 (2015-05-29)
Liquid fermentations of the fungus Stereum rameale (N° 2511) yielded extracts with antibacterial activity. The antibacterial activity reached its peak after 216 h of stirring. Bioassay-guided fractionation methods were employed for the isolation of the bioactive metabolites. Three known compounds
Nik Khairunissa Nik Abdullah Zawawi et al.
Bioorganic & medicinal chemistry, 23(13), 3119-3125 (2015-05-24)
A library of novel 2,5-disubtituted-1,3,4-oxadiazoles with benzimidazole backbone (3a-3r) was synthesized and evaluated for their potential as β-glucuronidase inhibitors. Several compounds such as 3a-3d, 3e-3j, 3l-3o, 3q and 3r showed excellent inhibitory potentials much better than the standard (IC50=48.4±1.25μM: d-saccharic
Barbara Parrino et al.
European journal of medicinal chemistry, 94, 367-377 (2015-03-18)
Three new ring systems, pyrido[2',3':3,4]pyrrolo[1,2-a]quinoxalines, pyrido[3',2':3,4]pyrrolo[1,2-a]quinoxalines and pyrido[2',3':5,6]pyrazino[2,1-a]isoindoles, were synthesized through an aza-substitution on the already active isoindolo-quinoxaline system and in particular in the position 7 or 4 of the isoindole moiety and in position 5 of the quinoxaline portion.
Priya P Netalkar et al.
European journal of medicinal chemistry, 79, 47-56 (2014-04-12)
Air and moisture stable coordination compounds of late first row transition metals, viz. Co(II), Ni(II), Cu(II) and Zn(II), with a newly designed ligand, 2-(2-benzo[d]thiazol-2-yl)hydrazono)propan-1-ol (LH), were prepared and successfully characterized using various spectro-analytical techniques. The molecular structures of the ligand
Papawee Suabjakyong et al.
PloS one, 10(3), e0122733-e0122733 (2015-03-27)
The basidiomycetous mushroom Phellinus igniarius (L.) Quel. has been used as traditional medicine in various Asian countries for many years. Although many reports exist on its anti-oxidative and anti-inflammatory activities and therapeutic effects against various diseases, our current knowledge of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service