Skip to Content
Merck
All Photos(1)

Key Documents

517070

Sigma-Aldrich

Nitrosylsulfuric acid solution

40 wt. % in sulfuric acid

Synonym(s):

Nitrosonium bisulfate, Nitrosonium hydrogen sulfate, Nitrosyl hydrogen sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€61.20

€61.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€61.20

About This Item

Linear Formula:
HO3SONO
CAS Number:
Molecular Weight:
127.08
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

€61.20


Check Cart for Availability

Request a Bulk Order

Quality Level

concentration

40 wt. % in sulfuric acid

density

1.865 g/mL at 25 °C

SMILES string

OS(=O)(=O)ON=O

InChI

1S/HNO5S/c2-1-6-7(3,4)5/h(H,3,4,5)

InChI key

VQTGUFBGYOIUFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service