Skip to Content
Merck
All Photos(1)

Key Documents

176303

Sigma-Aldrich

Nitronium tetrafluoroborate solution

0.5 M in sulfolane

Synonym(s):

Nitronium fluoroborate, Nitronium tetrafluoroboron, Nitryl fluoborate, Nitryl tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

200 ML
€391.00

€391.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 ML
€391.00

About This Item

Linear Formula:
NO2BF4
CAS Number:
Molecular Weight:
132.81
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.22

€391.00


Check Cart for Availability

Request a Bulk Order

concentration

0.5 M in sulfolane

density

1.268 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

O=[N+]=O.F[B-](F)(F)F

InChI

1S/BF4.NO2/c2-1(3,4)5;2-1-3/q-1;+1

InChI key

RGVZMPONLYFINH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Anologues of SCH 39166 as potent and selective dopamine D1 receptor antagonists
  • Anticonvulsant agent SB-406725A
  • Nitro-derivatives of chlorambucil conjugated to prasterone and pregnenolone as anticancer agents
  • Nano-sized cryptomelane particles for the formation of potentiometric potassium ion sensors
  • Nitro- and aminoindole derivatives
  • Hydroxynitroalkyl esters as affectors of metabolism of sulfate-reducing bacteria
  • Fluorescence probes to monitor the generation of nitrative stress in neurodegenerative disorders

Used as:
  • Oxidizing agent in high-power Li-ion battery technology for large-scale applications as required for plug-in hybrid vehicles

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service