Skip to Content
Merck
All Photos(2)

Key Documents

515205

Sigma-Aldrich

3-Indoleglyoxylyl chloride

98%

Synonym(s):

α-Oxoindole-3-acetyl chloride, 2-(3-Indolyl)-2-oxoacetyl chloride, NSC 128332

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6ClNO2
CAS Number:
Molecular Weight:
207.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

138 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)C(=O)c1c[nH]c2ccccc12

InChI

1S/C10H6ClNO2/c11-10(14)9(13)7-5-12-8-4-2-1-3-6(7)8/h1-5,12H

InChI key

FPEGGKCNMYDNMW-UHFFFAOYSA-N

Application

  • Reactant for preparation of dual IGF-1R/SRC inhibitors
  • Reactant for preparation of indolylglyoxylamides as a new class of antileishmanial agents
  • Reactant for preparation of glyoxyl analogs of indole phytoalexins as anticancer agents
  • Reactant for diastereoselective synthesis of heptacyclic core of dragmacidin E
  • Reactant for preparation of benzoylmethylpyrrolopyridinylethanedione as HIV-1 inhibitor
  • Reactant for preparation of fascaplysin-inspired diindolyls as selective inhibitors of CDK4/cyclin D1

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notes-Concerning a preparation of tryptamine.
Brutcher J, et al.
The Journal of Organic Chemistry, 23(1), 146-147 (1958)
A concise synthesis of an AHR endogenous ligand with the indolecarbonylthiazole skeleton.
Grzywacz P, et al.
Heterocycles, 60(5), 1219-1224 (2003)
Manabu Nakazono et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 19(1), 123-127 (2003-02-01)
The chemiluminescence (CL) intensities of various indole derivatives substituted with a glyoxylyl group at the 3-position and a hydroxyl group at the 5-position of the indole ring were compared upon the addition of H2O2 in alkaline media. The CL intensities

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service