Skip to Content
Merck
All Photos(1)

Key Documents

265284

Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzoyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€57.30

€57.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€57.30

About This Item

Linear Formula:
(CF3)2C6H3COCl
CAS Number:
Molecular Weight:
276.56
Beilstein:
2593440
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€57.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.435 (lit.)

density

1.526 g/mL at 25 °C (lit.)

functional group

acyl chloride
fluoro

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)C(Cl)=O

InChI

1S/C9H3ClF6O/c10-7(17)4-1-5(8(11,12)13)3-6(2-4)9(14,15)16/h1-3H

InChI key

WAKMMQSMEDJRRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Bis(trifluoromethyl)benzoyl chloride was used in the quantitative and selective assay for the measurement of low levels of clonidine in human plasma[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Girault et al.
Biomedical & environmental mass spectrometry, 17(6), 443-448 (1988-12-01)
A new quantitative and selective assay was developed for the measurement of low levels of clonidine in human plasma. The drug and the deuterated internal standard (2H4)clonidine are quantified by gas chromatography/negative ion chemical ionization mass spectrometry with methane as
D Premnath et al.
Interdisciplinary sciences, computational life sciences, 9(1), 130-139 (2016-01-05)
The commenced work deals with the synthesis, characterization and evaluation of biological activities of 4-amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one. The synthesis was done by the condensation of aromatic acid chlorides with 4-aminoantipyrine. The structures of synthesized derivatives were elucidated using IR, Mass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service