Skip to Content
Merck
All Photos(2)

Key Documents

512168

Sigma-Aldrich

3-Furanylboronic acid

≥95.0%

Synonym(s):

3-Furanboronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€87.40

€87.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€87.40

About This Item

Empirical Formula (Hill Notation):
C4H5BO3
CAS Number:
Molecular Weight:
111.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€87.40


Check Cart for Availability

Request a Bulk Order

Assay

≥95.0%

mp

139-144 °C (dec.) (lit.)

SMILES string

OB(O)c1ccoc1

InChI

1S/C4H5BO3/c6-5(7)4-1-2-8-3-4/h1-3,6-7H

InChI key

CYEFKCRAAGLNHW-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service