Skip to Content
Merck
All Photos(2)

Key Documents

464910

Sigma-Aldrich

2-Furanylboronic acid

≥95.0%

Synonym(s):

(5-Furanyl)boronic acid, 2-Furanboronic acid, 2-Furylboronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€92.10

€92.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€92.10

About This Item

Empirical Formula (Hill Notation):
C4H5BO3
CAS Number:
Molecular Weight:
111.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€92.10


Check Cart for Availability

Request a Bulk Order

Assay

≥95.0%

form

solid

mp

112 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

OB(O)c1ccco1

InChI

1S/C4H5BO3/c6-5(7)4-2-1-3-8-4/h1-3,6-7H

InChI key

PZJSZBJLOWMDRG-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Effective in key synthesis reactions like Knoevenagel condensation, thalidomide synthesis, and Suzuki coupling for sustainable chemical transformations.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service