Skip to Content
Merck
All Photos(2)

Key Documents

493457

Sigma-Aldrich

Trichloroacetic acid-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€1,760.00

€1,760.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€1,760.00

About This Item

Linear Formula:
Cl313CCO2H
CAS Number:
Molecular Weight:
164.38
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€1,760.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

form

solid

bp

196 °C (lit.)

mp

54-58 °C (lit.)

density

1.630 g/mL at 25 °C

mass shift

M+1

SMILES string

O[13C](=O)C(Cl)(Cl)Cl

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)/i1+1

InChI key

YNJBWRMUSHSURL-OUBTZVSYSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service