Skip to Content
Merck
All Photos(1)

Key Documents

485470

Sigma-Aldrich

Dichloroacetic acid-d2

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€1,200.00

€1,200.00


Check Cart for Availability

Request more information

Select a Size

Change View
10 G
€1,200.00

About This Item

Linear Formula:
Cl2CDCO2D
CAS Number:
Molecular Weight:
130.95
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€1,200.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Assay

99% (CP)

refractive index

n20/D 1.4663 (lit.)

bp

194 °C (lit.)

mp

9-11 °C (lit.)

density

1.587 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]OC(=O)C([2H])(Cl)Cl

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/i1D/hD

InChI key

JXTHNDFMNIQAHM-PFUFQJKNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Lact. - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A - STOT RE 2 Oral

Target Organs

Brain,Liver,Testes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maho Yagi-Utsumi et al.
Biophysical journal, 119(10), 2029-2038 (2020-11-04)
The characterization of residual structures persistent in unfolded proteins in concentrated denaturant solution is currently an important issue in studies of protein folding because the residual structure present, if any, in the unfolded state may form a folding initiation site

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service