Skip to Content
Merck
All Photos(1)

Key Documents

489921

Sigma-Aldrich

L-Alanine-3,3,3-d3

99 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€509.00

€509.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€509.00

About This Item

Linear Formula:
CD3CH(NH2)CO2H
CAS Number:
Molecular Weight:
92.11
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

€509.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % D

Assay

99% (CP)

form

solid

optical activity

[α]25/D +14.5°, c = 2 in 1 M HCl

mp

314.5 °C (dec.) (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])[C@H](N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i1D3

InChI key

QNAYBMKLOCPYGJ-VGCLAIIRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Zeng et al.
Journal of proteome research, 13(7), 3420-3431 (2014-05-24)
Hepatocellular carcinoma (HCC) is one of the most lethal malignancies. The lack of effective screening methods for early diagnosis has been a longstanding bottleneck to improve the survival rate. In the present study, a capillary electrophoresis-time-of-flight mass spectrometry (CE-TOF/MS)-based metabolomics
Toshiki Mochizuki et al.
Analytica chimica acta, 875, 73-82 (2015-05-06)
A novel triazine-type chiral derivatization reagent, i.e., (S)-2,5-dioxopyrrolidin-1-yl-1-(4,6-dimethoxy-1,3,5-triazin-2-yl) pyrrolidine-2-carboxylate (DMT-(S)-Pro-OSu), was developed for the highly sensitive and selective detection of chiral amines and amino acids by UPLC-MS/MS analysis. The enantiomers of amino acids were easily labeled with the reagents at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service