Skip to Content
Merck
All Photos(1)

Key Documents

486825

Sigma-Aldrich

L-Leucine-5,5,5-d3

99 atom % D

Synonym(s):

SILAC Amino Acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€604.00

€604.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€604.00

About This Item

Linear Formula:
CH3CH(CD3)CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
134.19
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

€604.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

solid

optical activity

[α]25/D +14.5°, c = 2 in 5 M HCl

technique(s)

protein expression: suitable

mp

>300 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i1D3/t4?,5-

InChI key

ROHFNLRQFUQHCH-LONBSJBQSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

L-leucine-5,5,5-d3 is an isotopic analogue of L-leucine.

Application

L-leucine-5,5,5-d3 is used in the quantitative determination of proteins using mass spectrometry and also used in the synthesis of (2S,4R)-(5,5,5-d3)leucine via stereospecific manner.
Used in the methodology of stable isotope tagging of epitopes (SITE) for identification of virus-induced peptides in vaccine development.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerald W Becker
Briefings in functional genomics & proteomics, 7(5), 371-382 (2008-12-25)
Straightforward methods for the introduction of stable isotopes into proteins with subsequent isolation and purification of the proteins will greatly aid the field of quantitative proteomics. Proteins containing amino acids with one or more of the stable isotopes of deuterium
Hugo D Meiring et al.
Molecular & cellular proteomics : MCP, 5(5), 902-913 (2006-01-25)
Identification of peptides presented in major histocompatibility complex (MHC) class I molecules after viral infection is of strategic importance for vaccine development. Until recently, mass spectrometric identification of virus-induced peptides was based on comparative analysis of peptide pools isolated from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service