Skip to Content
Merck
All Photos(1)

Key Documents

488771

Sigma-Aldrich

Diethyl malonate-1,2,3-13C3

99 atom % 13C

Synonym(s):

13C Labeled diethyl malonate, 13C Labeled diethyl malonic acid, Malonic acid diethyl ester-13C3

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€684.00

€684.00


Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
€684.00

About This Item

Linear Formula:
13CH2(13CO2C2H5)2
CAS Number:
Molecular Weight:
163.15
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

€684.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

refractive index

n20/D 1.414 (lit.)

bp

199 °C (lit.)

mp

−51-−50 °C (lit.)

density

1.074 g/mL at 25 °C

mass shift

M+3

SMILES string

CCO[13C](=O)[13CH2][13C](=O)OCC

InChI

1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3/i5+1,6+1,7+1

InChI key

IYXGSMUGOJNHAZ-SVFBATFISA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service