Skip to Content
Merck
All Photos(1)

Key Documents

488690

Sigma-Aldrich

Deuterium hydride

extent of labeling: 96 mol% DH, 98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DH
CAS Number:
Molecular Weight:
3.02
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

vapor density

0.07 (21 °C, vs air)

isotopic purity

98 atom % D

autoignition temp.

1060 °F

extent of labeling

96 mol% DH

bp

−252.8 °C (lit.)

mp

−259.2 °C (lit.)

mass shift

M+1

SMILES string

[H][2H]

InChI

1S/H2/h1H/i1+1

InChI key

UFHFLCQGNIYNRP-OUBTZVSYSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard products are packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressures at 21ºC are 20 psig for 1 L and 150 psig for 5 L. These pressures are slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951 or Z187224
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew C Singer et al.
PloS one, 9(9), e108621-e108621 (2014-09-26)
The concentration of eleven antibiotics (trimethoprim, oxytetracycline, ciprofloxacin, azithromycin, cefotaxime, doxycycline, sulfamethoxazole, erythromycin, clarithromycin, ofloxacin, norfloxacin), three decongestants (naphazoline, oxymetazoline, xylometazoline) and the antiviral drug oseltamivir's active metabolite, oseltamivir carboxylate (OC), were measured weekly at 21 locations within the River
Magdalena Sustkova-Fiserova et al.
Psychopharmacology, 231(14), 2899-2908 (2014-02-18)
Ghrelin, an orexigenic (appetite stimulating) peptide activates binding sites in the ventral tegmental area (a structure linked with the neural reward system) allowing it to participate in reward-seeking behavior. An increasing number of studies over the past few years have
Andrew J Ingram et al.
Journal of the American Chemical Society, 137(42), 13632-13646 (2015-10-08)
Aerobic oxidation of alcohols are catalyzed by the Pd-acetate compound [LPd(OAc)]2(OTf)2 (L = neocuproine = 2,9-dimethyl-1,10-phenanthroline) to form ketones and the release of hydrogen peroxide, but the latter rapidly undergoes disproportionation. We employ a series of kinetic and isotope labeling
Sui So et al.
Physical chemistry chemical physics : PCCP, 16(45), 24954-24964 (2014-10-18)
β-Hydroxyperoxyl radicals are formed during atmospheric oxidation of unsaturated volatile organic compounds such as isoprene. They are intermediates in the combustion of alcohols. In these environments the unimolecular isomerization and decomposition of β-hydroxyperoxyl radicals may be of importance, either through
Devalingam Mahalingam et al.
Autophagy, 10(8), 1403-1414 (2014-07-06)
We previously reported that inhibition of autophagy significantly augmented the anticancer activity of the histone deacetylase (HDAC) inhibitor vorinostat (VOR) through a cathepsin D-mediated mechanism. We thus conducted a first-in-human study to investigate the safety, preliminary efficacy, pharmacokinetics (PK), and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service