Skip to Content
Merck
All Photos(1)

Key Documents

488569

Sigma-Aldrich

Chlorpyrifos-(diethyl-d10)

99 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€2,380.00

€2,380.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
€2,380.00

About This Item

Empirical Formula (Hill Notation):
C9HD10Cl3NO3PS
CAS Number:
Molecular Weight:
360.65
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.12

€2,380.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % D

Quality Level

Assay

97% (CP)

mp

38-39 °C (lit.)

application(s)

agriculture
environmental

mass shift

M+10

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])OP(=S)(Oc1nc(Cl)c(Cl)cc1Cl)OC([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C9H11Cl3NO3PS/c1-3-14-17(18,15-4-2)16-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2

InChI key

SBPBAQFWLVIOKP-MWUKXHIBSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service