Skip to Content
Merck
All Photos(1)

Key Documents

487767

Sigma-Aldrich

Acetaldehyde-2,2,2-d3

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€455.00
5 G
€2,320.00
10 G
€4,190.00

€455.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€455.00
5 G
€2,320.00
10 G
€4,190.00

About This Item

Linear Formula:
CD3CHO
CAS Number:
Molecular Weight:
47.07
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€455.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

refractive index

n20/D 1.332 (lit.)

bp

21 °C (lit.)

mp

−125 °C (lit.)

density

0.838 g/mL at 25 °C

mass shift

M+3

storage temp.

−20°C

SMILES string

[H]C(=O)C([2H])([2H])[2H]

InChI

1S/C2H4O/c1-2-3/h2H,1H3/i1D3

InChI key

IKHGUXGNUITLKF-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-29.2 °F - closed cup

Flash Point(C)

-34 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy M Green et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(47), 12372-12377 (2017-11-08)
Ozonolysis of alkenes, an important nonphotolytic source of hydroxyl (OH) radicals in the atmosphere, proceeds through unimolecular decay of Criegee intermediates. Here, we report a large kinetic isotope effect associated with the rate-limiting hydrogen-transfer step that releases OH radicals for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service