Skip to Content
Merck
All Photos(1)

Key Documents

485225

Sigma-Aldrich

γ-Butyrolactone-d6

98 atom % D

Synonym(s):

Dihydro-d2-2(3H)-furanone-3,3,4,5-d4

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€2,800.00

€2,800.00


Check Cart for Availability

Request more information

Select a Size

Change View
5 G
€2,800.00

About This Item

Empirical Formula (Hill Notation):
C4D6O2
CAS Number:
Molecular Weight:
92.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

€2,800.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

204-205 °C (lit.)

mp

−45 °C (lit.)

density

1.197 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]C1([2H])OC(=O)C([2H])([2H])C1([2H])[2H]

InChI

1S/C4H6O2/c5-4-2-1-3-6-4/h1-3H2/i1D2,2D2,3D2

InChI key

YEJRWHAVMIAJKC-NMFSSPJFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service